Goggles, uvex u-sonic
Catalog # 111-1706
Supplier: UVEX
Specifications
- Type:Anti-UV, anti-scratch, anti-fog, chemical resistant
- Colour:Grey/green
- Lenses:Clear PC
- Pk:1
Specifications
About this item
The ergonomic design with flexible soft component means the uvex u-sonic fits perfectly to every face its low weight and low pressure fit make them very comfortable to wear.
- Combines hard and soft components which adapt to the wearer’s face creating soft, effective seal around the eyes
- Fully adjustable, high quality head strap offers pressure-free fit
- Prescription insert possible
- Compact size and soft materials aid compatibility with respirators and other PPE items
- Innovative ventilation system ensures pleasant comfortable interior environment
- Large panorama lens is interchangeable
Can be used with add-on lenses. The integrated magnet technology makes it easy to swap out the add-on lens.
Weight: 69 g
Certifications: EN 166, EN 170, EN 172