Anti-CD10 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SN5c]
Supplier: Tonbo Biosciences
Certificates
About this item
The SN5c antibody is specific for human CD10, a 100 kDa type II transmembrane protein also known as Common Acute Lymphoblastic Leukemia Antigen (CALLA). In normal cells, it is expressed on early B and T lymphoid precursors, neutrophils and on various epithelia. CD10 is also expressed on several lymphoma cells including Burkitt�s, acute lymphoblastic, and follicular germinal center lymphomas. CD10 functions as an endopeptidase and is involved in B cell development, as well as a mediator of neutrophil inflammatory responses.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: SN5c
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 60-0108-T500
- 60-0108-T100
- 60-0108-T025
- Antigen symbol:
- MME
- MME
- MME
- Antigen name:
- Membrane metallo-endopeptidase
- Membrane metallo-endopeptidase
- Membrane metallo-endopeptidase
- Antigen synonyms:
- NEP|CD10|CALLA|SFE
- NEP|CD10|CALLA|SFE
- NEP|CD10|CALLA|SFE
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)-Cy7®
- PE (Phycoerythrin)-Cy7®
- PE (Phycoerythrin)-Cy7®
- Clone:
- SN5c
- SN5c
- SN5c
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4311
- 4311
- 4311
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.