Order Entry
Germany
ContactUsLinkComponent
Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex))
Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 67-4724-U100
Supplier:  Tonbo Biosciences
Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 67-4724-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    C1.18.4
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Cross adsorption:
    No
  • Storage temperature:
    Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Pk:
    100 µG

Specifications

About this item

The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: