Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 61D3]
Supplier: Tonbo Biosciences
Certificates
About this item
The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 20-0149-T100
- 20-0149-T500
- 20-0149-T025
- Antigen symbol:
- CD14
- CD14
- CD14
- Antigen name:
- CD14 Molecule
- CD14 Molecule
- CD14 Molecule
- Antigen synonyms:
- monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- 61D3
- 61D3
- 61D3
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 929
- 929
- 929
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.