KAPA™ Library Quantification kits
Supplier: KAPA BIOSYSTEMS
Certificates
About this item
Accurate quantification of Next Generation Sequencing (NGS) libraries is crucial for reliable clonal amplification via either emulsion or bridge PCR. Underestimation results in non-clonality, while overestimation leads to inefficiency.
KAPA™ Library Quantification Kits combine a quality-controlled set of DNA standards with the performance of KAPA™ SYBR® FAST qPCR reagents to provide a rapid, sensitive, and reliable method for quantifying amplifiable molecules in NGS libraries.
Delivery: Kits contain primers, DNA standards, and Real-Time PCR reagents specific for both DNA sequencing platform (Illumina GA, Illumina HiSeq, MiSeq, Roche 454 GS FLX, Roche 454 GS Titanium, Roche 454 GS Junior, ABI SOLiD, Ion Torrent PGM) and qPCR instrument (universal, ABI Prism®, Bio-Rad iCycler™, Roche LightCycler®480).