Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microplates
PCR Plates
PCR plates, 96-well, with half skirt, Multiply®
PCR plates, 96-well, with half skirt, Multiply®
PCR plates, 96-well, with half skirt, Multiply®
Catalog # SARS72.1979
Supplier:
SARSTEDT
Expand image
PCR plates, 96-well, with half skirt, Multiply®
Catalog # SARS72.1979
Supplier:
SARSTEDT
Specifications
No. of wells:
96
Profile:
Standard
Skirt:
Semi-skirted, raised rim
Well colour:
Clear
Barcoded:
No
Pk:
100
Specifications
About this item
Product Details
Product Details