Anti-FRA2 Antibody
Catalog # BSBTPB9591
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:FOS-like antigen 2
- Antigen symbol:FOSL2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2355
- Antigen synonyms:FRA2
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Fos-related antigen 2(FOSL2) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: FRA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat