Anti-MCP1 Antibody
BSBTPB9570
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 2
- Antigen symbol:CCL2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6347
- Antigen synonyms:HC11|MCAF|SMC-CF|GDCF-2|HSMCR30|SCYA2|MCP1|MCP-1
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for C-C motif chemokine 2(CCL2) detection. Tested with WB, ELISA in Human.
Type: Primary
Antigen: MCP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human