X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Catalog # 730-1498
Supplier: VWR Peqlab
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:408.63 g/mol
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666