Order Entry
Germany
ContactUsLinkComponent
VWR®, Protein marker III (pre-stained), peqGOLD
VWR®, Protein marker III (pre-stained), peqGOLD
Catalog # 27-1110
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Protein marker III (pre-stained), peqGOLD
Catalog # 27-1110
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Protein Marker III (pre-stained, ready to use), 2×250 µl, sufficient for 50 lanes

Specifications

About this item

Protein Marker III for monitoring protein separation, transfer efficiency and size determination.

  • Mix of six highly purified proteins
  • Range: 20 to 120 kDa
  • Concentration: 200 ng/µl each protein
  • Each protein covalently pre-stained with a blue colouring
  • Ready to use for a fast, convenient workflow