ContactUsLinkComponent
Peqlab peqGOLD, Protein Marker II (Unstained)
Catalog # 27-2010
Supplier:  VWR Chemicals
CAS Number:  
undefined
Peqlab peqGOLD, Protein Marker II (Unstained)
Catalog # 27-2010
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Protein Marker II (unstained, ready to use), 2×250 µl, sufficient for 50 lanes
Specifications
About this item
Protein Marker II for accurate molecular weight determination of proteins by SDS-PAGE.
  • Mix of 14 recombinant proteins
  • Range: 10 to 200 kDa
  • Each protein with Strep-Tag® II sequence (WSHPQFEK)
  • Highlighted 50 kDa band for easy orientation
  • Ready to use for a fast, convenient workflow