Drape sheets, Foliodrape® Comfort
Foliodrape® Protect drape sheets, sterile, individually wrapped, 500×500 mm
Supplier: HARTMANN PAUL
Jump to:
Product Details & Documents
These surgical drape sheets are made from a soft three layer material. For sterile draping of patients or equipment in the operating theatre.
- Impermeable to moisture and bacteria
- Abrasion resistant, extremely low linting and highly conformable
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Foliodrape® Protect drape sheets, sterile, individually wrapped
- L×W:500×500 mm