Precision alcohol meters
Supplier: Amarell
Certificates
About this item
Alcoholmeters of commercial grade.
- With or without thermometer
- Paper scales with well visible print
- Thermometer inside the hydrometer body
- Shot weighted body
- Filling: Hg or Paraffine Oil (Pet)
Certifications: EU Directive 847/2012: Mercury products must not be sold in EU countries after April 10th, 2014 .