Film frame rings for wafers
: ENTEGRIS
Plastic film frame rings provide a cost-effective and light weight alternative to expensive, heavy, metal frames to handle wafers during assembly operations and can be used to store and ship finished wafers.
- Film frames rings are available for 300, 200 and 150 mm wafers