Orders LinkContactUsLinkComponent
Digital timer, four channels
Digital timer, four channels
Digital timer, 4 channels, White/grey, 65×25×85 mm
Catalog # MOIN437/2E
Supplier:   MOINEAU
Digital timer, four channels
Catalog # MOIN437/2E
Supplier:  MOINEAU
Digital timer, four channels
Digital timer, four channels
Digital timer, 4 channels, White/grey, 65×25×85 mm
Price
104,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This digital timer offers four measuring channels (1× count up, 3× count down).
  • Timing capacity: 20 hours (1/100 seconds)
  • Independent or simultaneous measurements
  • With clock
  • Three alarms, three memories
Delivery information: Supplied with stand, magnet and two LR44 batteries.
Specifications
  • Colour:
    White/grey
  • Description:
    Digital timer, 4 channels
  • W×D×H:
    65×25×85 mm