Order Entry
Germany
ContactUsLinkComponent
Anti-SH2D4A Polyclonal Antibody Pair
Anti-SH2D4A Polyclonal Antibody Pair
Catalog # ABNOH00063898-PW1
Supplier:  Abnova
Anti-SH2D4A Polyclonal Antibody Pair
Catalog # ABNOH00063898-PW1
Supplier:  Abnova

Specifications

  • Antigen name:
    SH2 domain containing 4A
  • Antigen symbol:
    SH2D4A
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    63898
  • Antigen synonyms:
    SIAH1A|PPP1R38|SH2A
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-SH2D4A (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-SH2D4A (50 µg)

Type:
Antigen: SH2D4A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human