Order Entry
Germany
ContactUsLinkComponent
 
Anti-MBD5 Antibody Pair
  ABNOH00055777-AP11
 :  Abnova
Anti-MBD5 Antibody Pair
  ABNOH00055777-AP11
 :  Abnova

 

  • Antigen name:
    Methyl-CpG binding domain 5 DNA glycosylase
  • Antigen symbol:
    MBD5
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    55777
  • Antigen synonyms:
    MRD1
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human MBD5.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-MBD5 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-MBD5, IgG1 kappa (20 µg);

Type:
Antigen: MBD5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human