Protein digestion kits, trypsin kits, SMART Digest™
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
Thermo Scientific™ SMART Digest™ Trypsin Kits are designed to provide a simple to use, fast, highly reproducible and robust process for digestion of proteins in a high throughput format, meeting the needs of biopharmaceutical, bioanalytical and QA/QC environments.
- Simple process with significantly fewer steps compared to in-solution digests
- Reduced sample preparation time, typically taking <60 minutes (dependant on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours
- Higher reproducibility and greater sensitivity than other methods
- Easily automated for high throughput processing of samples
Delivery: Available as a stand alone kit, or with optional filter plate or SOLAµ™ SPE plate. Bulk resin (-B) and magnetic bulk resin (-MB) are also available separately.