Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
SYBR® Green I
SYBR® Green I nucleic acid stain
SYBR® Green I nucleic acid stain
SYBR® Green I nucleic acid stain
Supplier:
LONZA
Certificates
Expand image
SYBR® Green I nucleic acid stain
Supplier:
LONZA
About this item
CAS Number:
163795-75-3
:
Formula:
C₃₂H₃₇N₄S
MDL Number:
MFCD00284715
:
509.73 g/mol