ContactUsLinkComponent
Hair analysis set
Hair analysis set
Catalog # SUREHA
Supplier:  SURESCREEN
undefined
Hair analysis set
Catalog # SUREHA
Supplier:  SURESCREEN
Specifications
  • Description:
    Hair analysis for drugs of abuse
  • Pk:
    1
Specifications
About this item
Hair is a medium designed to analyse drug abuse over a period of three months. As the hair grows it records all the drugs that are in the body. Hair is easy to collect and cannot be altered.
  • Standard hair test checks for the presence
  • Amphetamines
  • Cannabis (THC)
  • Cocaine
  • Methamphetamines
  • Opiates
  • PCP
Standard hair test checks for the presence of: Amphetamines; Cannabis (THC); Cocaine; Methamphetamines; Opiates; PCP.