Tubing, polyethylene
Supplier: BURKLE
New Product
Certificates
About this item
PE, translucent.
- Temperature resistant from -40 to +95 °C
- Hardness Shore A 80 ±3
- Suitable for food
Recognised as safe physiologically, resistant against (non-oxidizing) acids, caustic solutions, salts and a number of solvents, pressure resistant, flexible, impact resistant, light, resistant to compression, up to + 80 °C deformation resistant.