Films for agarose gels, GelBond®
Supplier: LONZA
Certificates
About this item
GelBond® film is a transparent, flexible polyester film designed to support agarose gels. Gels cast on GelBond® film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures. GelBond® Film is available either as precut sheets or rolls.
- Agarose gels retain their original dimensions during staining and after drying
- Gels, particularly thin ones, are easier to handle during staining, destaining, and drying when supported
- Gel orientation can be recorded directly on the GelBond® PAG film prior to casting
NOTE: Polyester films will not transmit light of less than 310 nm, and will fluoresce at higher wavelengths.