Films for acrylamide gels, GelBond® PAG
Supplier: LONZA
Certificates
About this item
GelBond® PAG film is a transparent, flexible polyester film designed to support polyacrylamide or MDE™ gels. Gels remain permanently attached to the film through electrophoresis and all subsequent fixing, staining, destaining and drying procedures.
- Polyacrylamide gels retain their original dimensions during staining and after drying
- Gels, particularly thin ones, are easier to handle during staining, destaining, and drying when supported
- Gel orientation can be recorded directly on the GelBond® PAG film prior to casting