Quanti-Cult® System
Quanti-Cult® products contain Quality Control organisms in a ready-to-use (<100 CFU) format, saving time, money, and reducing procedure time from 2 to 3 days to 15 minutes. Preserved American Type Culture Collection (ATCC®) strains are provided as a film inside the vial cap on a plastic vial, ready for rehydration and use in QC procedures. The Quanti-Cult® System is available in two formats: Quanti-Cult® and Quanti-Cult Plus®. Quanti-Cult® delivers <100 CFUs per single 0.3 ml inoculum. Quanti-Cult Plus® delivers <100 CFUs as ten 0,1 ml inocula, providing scalability when validation protocols mandate the need for multiple assay runs.
- No serial dilution steps eliminate long growth periods and stock culture maintenance
- Transferable labels allow fast and accurate documentation
- No need for sharps, pipettes, or forceps
- QC organisms are produced using patented Gentle Gel Preservation Process (no lyophilisation), standardizing CFUs among all personnel performing QC procedures, reducing errors during use and enhancing organism recovery
- Organisms are stable (18 month shelf-life from date of manufacture), allowing you flexibility with your time
- Meets USP requirements