Protease inhibitor cocktail, Yeast/Fungal ProteaseArrest™
Catalog # GENO786-333
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 mL
- Description:Yeast/Fungal ProteaseArrest™ (100X)
Specifications
About this item
A broad range, 100X concentrated, ready-to-use protease inhibitor cocktail, Yeast/ Fungal ProteaseArrest™ inhibits yeast and fungal serine, cysteine and metalloproteases.
Applications include inhibition of protease activity in protein preparations of yeast and fungal lysates, protection of proteins from proteolysis in such applications as electrophoresis, purification, storage, assays, and other applications.