ContactUsLinkComponent
DNA ladder, 100 bp, DNAmark™
DNA ladder, 100 bp, DNAmark™
Catalog # GENO786-855
Supplier:  G-Biosciences
CAS Number:  
undefined
DNA ladder, 100 bp, DNAmark™
Catalog # GENO786-855
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    DNAmark™ 100 bp ladder, 50 µg
  • Seq:
    03
Specifications
About this item
DNAmark™ 100 bp ladder consists of 11 fragments ranging from 100 to 1500 base pairs (bp). The 500 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500 bp are 40 ng. The 500 bp fragment is 100 ng.
  • Pre-mixed with loading dye and ready to use