Specifications
- Antibody type:Primary
- Antigen name:Gastrin
- Antigen symbol:GAST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:Human Gastrin peptide (76-85 aa)
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Gastrin component III|Gastrin-34|GAST_HUMAN|Gastrin-17|Gastrin-6|Gastrin component I|Gastrin component II|Gastrin precursor
- Immunogen:Synthetic human Gastrin peptide (76-85 aa) conjugated to BSA.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Human Gastrin is a 101 amino acid hormone produced by G cells of the duodenum, stomach and pancreas. It stimulates secretion of hydrochloric acid by parietal cells of the stomach. Gastrin is also secreted into bloodstream.
Antibody recognizes Gastrin in multiple species and in native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Gastrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Gastrin peptide (76-85 aa)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat