Order Entry
Germany
ContactUsLinkComponent
Anti-NT3 Rabbit Polyclonal Antibody
Anti-NT3 Rabbit Polyclonal Antibody
Catalog # BSENR-175-20
Supplier:  Biosensis
undefined
Anti-NT3 Rabbit Polyclonal Antibody
Catalog # BSENR-175-20
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurotrophin-3
  • Antigen symbol:
    NT3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Chicken,
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    aa: 139-149), Mouse and rat NT3 protein
  • Cross adsorption:
    No
  • Neutralisation:
    Yes
  • Format:
    Lyophilized
  • Immunogen:
    A synthetic peptide (YAEHKSHRGEY) as part of human (aa: 139-149), mouse and rat NT3 protein conjugated to BSA has been used as the immunogen.
  • Size:
    20 µg
  • Pk:
    20 µG

Specifications

About this item

NT3 is a member of the neurotrophin family, that controls survival and differentiation of visceral and proprioceptive sensory neurons. NT3 is closely related to both NGF and BDNF. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NT3-deficient mice generated by gene targeting display sevvere movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family.

A cross reactivity of less than 0.1% to mouse NGF, recombinant human BDNF and NT4/5 has been shown by dot blot.
Application Information:
IHC, ELISA, WB, inhibition of biological activity. A concentration of 1-10 µg/ml is recommended for IHC, ELISA, WB and inhibition of biological activity in vitro; 2-10 µg/ml (ED50) for in vivo use. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa: 139-149), Mouse and rat NT3 protein
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Chicken, Human