Specifications
- Antibody type:Primary
- Antigen symbol:rh NT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Epitope:Multiple points of reactivity likely because of the polyclonal nature of the antibody
- Cross adsorption:No
- Neutralisation:Yes
- Format:Lyophilized
- Antigen synonyms:Neurotrophin-5|NTF4,
- Immunogen:Recombinant human NT4
- Size:500 µg
- Pk:500 µG
Specifications
About this item
FUNCTION: Target-derived survival factor for peripheral sensory sympathetic neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Highest levels in prostate, lower levels in thymus, placenta, and skeletal muscle. Expressed in embryonic and adult tissues. SIMILARITY: Belongs to the NGF-beta family.
Less than 1% cross-reactivity against NGF, recombinant human BDNF and 5% to NT3 has been shown by dot blot.
Application Information:
IHC, ELISA (1 site), Western Blot, ICC, inhibition of biological activity in vitro/in vivo. Recommended to be used at a concentration of 1-10 µg/ml for immunohistochemistry, ELISA, ICC and Western blot and inhibition of biological activity in vitro. Use neat for in vivo studies at 2-10 µg/ml (ED50). Note that the concentration of NT4 is generally low in most tissues nevertheless, neonatal testes of rat can be used as a good positive control. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: rh NT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human, Mouse, Monkey