Order Entry
Germany
ContactUsLinkComponent
Anti-rh NT4 Sheep Polyclonal Antibody
Anti-rh NT4 Sheep Polyclonal Antibody
Catalog # BSENS-059-500
Supplier:  Biosensis
undefined
Anti-rh NT4 Sheep Polyclonal Antibody
Catalog # BSENS-059-500
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    rh NT4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Multiple points of reactivity likely because of the polyclonal nature of the antibody
  • Cross adsorption:
    No
  • Neutralisation:
    Yes
  • Format:
    Lyophilized
  • Antigen synonyms:
    Neurotrophin-5|NTF4,
  • Immunogen:
    Recombinant human NT4
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

FUNCTION: Target-derived survival factor for peripheral sensory sympathetic neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Highest levels in prostate, lower levels in thymus, placenta, and skeletal muscle. Expressed in embryonic and adult tissues. SIMILARITY: Belongs to the NGF-beta family.

Less than 1% cross-reactivity against NGF, recombinant human BDNF and 5% to NT3 has been shown by dot blot.
Application Information:
IHC, ELISA (1 site), Western Blot, inhibition of biological activity in vitro/in vivo. Recommended to be used at a concentration of 2-10 µg/ml for immunohistochemistry, ELISA and Western blot and inhibition of biological activity in vitro. Use neat for in vivo studies at 2-10 µg/ml (ED50). Note that the concentration of NT4 is generally low in most tissues nevertheless, neonatal testes of rat can be used as a good positive control. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: rh NT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Sheep
Isotype: IgG
Reactivity: Rat, Human, Mouse, Monkey