Latex agglutination test, Wellcolex®
Supplier: THERMO FISHER DIAGNOSTICS
Certificates
About this item
Wellcolex® Rapid Latex Agglutination test kits are designed for rapid and accurate identification of enteric bacteria, such as Salmonella, Shigella and E.coli. With its high positive and negative predictive values (PPV >98,7%) and results in less than three minutes, laboratories can use this rapid screening test to eliminate Salmonella-negative samples from further testing, reducing the number of samples requiring confirmatory testing.
- Easy to use
- Save up to two days' testing time over traditional screening procedures