Viabank™ Bacterial Storage Beads
Supplier: MEDICAL WIRE EQUIPMENT
Certificates
About this item
Viabank™ bacterial storage beads for long term storage of microorganisms at low temperatures.
- Twenty glass beads per vial
- Coloured caps match colour of beads
- Available in multi coloured packs of red, yellow, green and blue or singular packs of each colour
- Individual beads can be removed while frozen at any time for establishing fresh cultures