Tubings, PFA, high purity, PureBond® 4200
Supplier: ENTEGRIS
Certificates
About this item
Made of PFA, DuPont 440.
Permanent serialized laser marking is a nondestructive and noncontaminating method to identify tube material and Ø. It also allows for 100% traceability to a specific material lot, production date and production inspection record.