ContactUsLinkComponent
Detachment reagents, macrophage detachment solution DXF
Detachment reagents, macrophage detachment solution DXF
Catalog # C-41330
Supplier:  PROMOCELL
CAS Number:  
undefined
Detachment reagents, macrophage detachment solution DXF
Catalog # C-41330
Supplier:  PROMOCELL
CAS Number:  
Specifications
  • Pk:
    250 mL
  • Description:
    Macrophage Detachment Solution DXF, 250 ml
Specifications
About this item
Macrophage Detachment Solution DXF is a non-enzymatic reagent especially optimised for the gentle detachment of strongly adherent macrophages.