Order Entry
Germany
ContactUsLinkComponent
Anti-MLXIPL Mouse Monoclonal Antibody [clone: 5B2]
Anti-MLXIPL Mouse Monoclonal Antibody [clone: 5B2]
Catalog # ABNOH00051085-M01
Supplier:  Abnova
Anti-MLXIPL Mouse Monoclonal Antibody [clone: 5B2]
Catalog # ABNOH00051085-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MLX interacting Protein-like
  • Antigen symbol:
    MLXIPL
  • Clonality:
    Monoclonal
  • Clone:
    5B2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    51085
  • Antigen synonyms:
    CHREBP|WBSCR14|WS-bHLH|bHLHd14|MIO|MONDOB
  • Amino acid number:
    603 to 700
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    PPAPSGSERRLSGDLSSMPGPGTLSVRVSPPQPILSRGRPDSNKTENRRITHISAEQKRRFNIKLGFDTLHGLVSTLSAQPSLKVSKATTLQKTAEYI
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    MLXIPL (NP_116569.1, 603 a.a. ~ 700 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a partial recombinant MLXIPL.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: MLXIPL
Clonality: Monoclonal
Clone: 5B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: