Order Entry
Germany
ContactUsLinkComponent
Anti-PRKAA1 Rabbit Polyclonal Antibody
Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25393
Supplier:  Abnova
Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25393
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 1
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5562
  • Antigen synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Modification:
    phospho-Thr174
  • Storage buffer:
    PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
  • Sequence:
    L-R-Tp-S-C
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide corresponding to residues surrounding T174 of human PRKAA1.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKAA1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat