Anti-CDH11 Mouse Monoclonal Antibody [clone: 2C67]
ABNOH00001009-M25
: Abnova
- Antibody type:Primary
- Antigen name:Cadherin 11
- Antigen symbol:CDH11
- Clonality:Monoclonal
- Clone:2C67
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1009
- Antigen synonyms:OB|Cadherin 11|OSF-4|OB-Cadherin (osteoblast)|OB-Cadherin|Cadherin-11|CDHOB|CAD11|osteoblast Cadherin|type 2
- Amino acid number:54 to 612
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CDH11 (P55287, 54 a.a. ~ 612 a.a) partial recombinant protein with Fc tag at C terminal. MW of the Fc tag alone is 50 KDa.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against a partial recombinant CDH11.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CDH11
Clonality: Monoclonal
Clone: 2C67
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human