Anti-MECP2 Rabbit Polyclonal Antibody
ABNOPAB0646
: Abnova
- Antibody type:Primary
- Antigen name:Methyl CpG binding protein 2
- Antigen symbol:MECP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Format:Liquid
- Gene ID:17257
- Antigen synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Storage buffer:PBS (0,05% sodium azide, 0,05% ProCl300)
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of mouse MeCP2.
- Purification:Affinity purification
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against synthetic peptide of Mecp2.
- Recommended Dilutions: ChIP: 5 μg/ChIP; ELISA: 1:1000; Western Blot: 1:1000
Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse