Anti-MECP2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB0170
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Methyl CpG binding protein 2
- Antigen symbol:MECP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4204
- Antigen synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Amino acid number:11 to 25 and 181 to 195
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides corresponding to amino acids 11-25 and 181-195 of human MECP2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MECP2.
Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human