Order Entry
Germany
ContactUsLinkComponent
 
Anti-PRKAA1 Rabbit Polyclonal Antibody
  ABNOPAB14475
 :  Abnova
Anti-PRKAA1 Rabbit Polyclonal Antibody
  ABNOPAB14475
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 1
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5562
  • Antigen synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Storage buffer:
    Tris-citrate/phosphate, pH 7-8 (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    A synthetic peptide corresponding to the portion encoded within exon 8 of human PRKAA1.
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody raised against synthetic peptide of PRKAA1.

Recommended Dilutions: Western Blot: 0.2-2 ?g/ml; Immunoprecipitation: 2-8 ?g/mg)

Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Rat