Anti-GDF15 Mouse Monoclonal Antibody [clone: 7C12.B3.F2]
Catalog # ABNOMAB8162
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Growth differentiation factor 15
- Antigen symbol:GDF15
- Clonality:Monoclonal
- Clone:7C12.B3.F2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9518
- Antigen synonyms:MIC-1|GDF-15|NAG-1|PTGFB|PDF|PLAB|MIC1
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus end of human GDF15 .
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of GDF15 .
Recommended Dilutions: ELISA: 1:150000; Western Blot: 1:1000
Type: Primary
Antigen: GDF15
Clonality: Monoclonal
Clone: 7C12.B3.F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human