Anti-DOCK4 Rabbit Polyclonal Antibody
ABNOPAB15726
: Abnova
- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 4
- Antigen symbol:DOCK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:238130
- Antigen synonyms:C030023J22|AF263288|5330406C03|6330411N01Rik|mKIAA0716
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Sequence:CLSPRDRPCSAIYPTPVEPSQRMLFNHIGDGALPRSDPNLSAPEKAVNPTPSSWSLDSGKEAKNMSDSGKLISPPVPPRPTQTASPARHTTSVSPSPAGRSPLKGSVQSFTPSPVEYNSPGLSSNSPVLSGSYSSGISSLSRCSTSETSGFENQANEQSVPVPVPVPVPVPVPSFSGSEEPVRKESKTPPPYSVYERTLRRPVPLPHSLSIPVTSEPPALPPKPLAARSSHLENGTRRTEPGPRPRPLPRKVSQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 254 mouse Dock4.
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against partial recombinant Dock4.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: DOCK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse