Anti-CD1 Mouse Monoclonal Antibody [clone: 76-7-4]
Catalog # ABNOMAB6216
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD1
- Clonality:Monoclonal
- Clone:76-7-4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Pig
- Cross adsorption:No
- Format:Liquid
- Gene ID:396800
- Storage buffer:100 mM BBS, pH 8,2
- Storage temperature:Store at 4 °C.
- Immunogen:dd Miniature swine thymocytes.
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against CD1.
Type: Primary
Antigen: CD1
Clonality: Monoclonal
Clone: 76-7-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pig