Order Entry
Germany
ContactUsLinkComponent
Anti-Grin1 Mouse Monoclonal Antibody [clone: R1JHL]
Anti-Grin1 Mouse Monoclonal Antibody [clone: R1JHL]
Catalog # ABNOMAB6237
Supplier:  Abnova
Anti-Grin1 Mouse Monoclonal Antibody [clone: R1JHL]
Catalog # ABNOMAB6237
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 1
  • Antigen symbol:
    GRIN1
  • Clonality:
    Monoclonal
  • Clone:
    R1JHL
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Form:
    Culture Supernatant
  • Gene ID:
    24408
  • Antigen synonyms:
    NMDAR1|GluN1|NR1
  • Amino acid number:
    1 to 564
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 50 ?l PBS, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 1-564 of rat Grin1.
  • Size:
    15 μg
  • Pk:
    15 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant Grin1.

Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 3 ?g/200 ?g lysate)

Type: Primary
Antigen: Grin1
Clonality: Monoclonal
Clone: R1JHL
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat