Anti-HIVEP2 Goat Polyclonal Antibody
ABNOPAB17829
: Abnova
- Antibody type:Primary
- Antigen name:Immunodeficiency virus type I enhancer binding Protein 2
- Antigen symbol:HIVEP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Cross adsorption:No
- Format:Liquid
- Gene ID:3097
- Antigen synonyms:MIBP1|HIV-EP2|SHN2|ZNF40B|ZAS2|MBP-2
- Storage buffer:0,5 mg/mL Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
- Sequence:C-TEESKDPSSEKSQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human HIVEP2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of HIVEP2.
Recommended Dilutions: ELISA: 1:1000
Type: Primary
Antigen: HIVEP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: