Order Entry
Germany
ContactUsLinkComponent
Anti-PRKAG1 Rabbit Polyclonal Antibody
Anti-PRKAG1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19812
Supplier:  Abnova
Anti-PRKAG1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19812
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 1
  • Antigen symbol:
    PRKAG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Serum
  • Gene ID:
    5571
  • Antigen synonyms:
    5'-AMP-activated protein kinase subunit gamma-1|AMPKG3|AMPK gamma3|PRKAG3|AMPK- gamma3|5'-AMP-activated protein kinase subunit gamma-2|AMPK- gamma1|AMPK subunit gamma-1|AMPKg|AMPK subunit gamma-3|H91620p|PRKAG2|AMPK- gamma2|AMPK subunit gamma-2|PRKAG1|AMPK gamma2|AMPK gamma1|5'-AMP-activated protein kinase subunit gamma-3
  • Storage buffer:
    0,5% BSA, 0,02% sodium azide
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 15 amino acids at internal region of human PRKAG1.
  • Purification:
    Peptide affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of PRKAG1.

Recommended Dilutions: ELISA: 1:5000-1:20000; Immunohistochemistry: 1:100-500

Type: Primary
Antigen: PRKAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat