Order Entry
Germany
ContactUsLinkComponent
Anti-PRKAA2 Rabbit Polyclonal Antibody
Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19814
Supplier:  Abnova
Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19814
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 2
  • Antigen symbol:
    PRKAA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Serum
  • Gene ID:
    5563
  • Antigen synonyms:
    AMPKa2|AMPK|AMPK2|PRKAA
  • Storage buffer:
    0,02% sodium azide
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 16 amino acids at internal region of human PRKAA2.
  • Purification:
    Protein A purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of PRKAA2.

Recommended Dilutions: ELISA: 1:20000-80000; Western Blot: 1:200-1000

Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat