Order Entry
Germany
ContactUsLinkComponent
Anti-RER1 Goat Polyclonal Antibody
Anti-RER1 Goat Polyclonal Antibody
  ABNOPAB7598
 :  Abnova
Anti-RER1 Goat Polyclonal Antibody
  ABNOPAB7598
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    retention in endoplasmic reticulum sorting receptor 1
  • Antigen symbol:
    RER1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    11079
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    C-HGKRRYRGKEDAGK
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human RER1.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Goat polyclonal antibody raised against synthetic peptide of RER1.

Recommended Dilutions: ELISA: 1:128000

Type: Primary
Antigen: RER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: