Anti-Human IgG Monoclonal Antibody (Unconjugated)
Catalog # ABNOPAB9323
Supplier: Abnova
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Liquid
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human IgG whole molecule
- Purification:solid phase adsorption
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Sheep anti-human IgG recognizes human IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host:
Isotype:
Reactivity: Human