About this item
The O323 antibody reacts with human CD27 (TNFRSF7), a cell surface homodimer of 55 kDa subunits, which provides co-stimulatory signaling in support of the T cell (TCR) and B cell (BCR) receptors. By comparison with CD28, whose TCR co-stimulatory signal can trigger cell proliferation, CD27 signaling appears to promote cell survival and differentiation to effector / memory stages. Also in contrast with CD28, the CD27 receptor may be shed following interaction with its ligand CD70, which is typically expressed on activated dendritic cells, T cells and B cells. With respect to B cells, CD27 is considered to be a phenotypic marker for memory B cells. CD27 has been included within a group of phenotypic markers for identifying human B regulatory cells (Bregs), a cell type proposed to regulate CD4+ T cell proliferation and Foxp3 / CTLA-4 expression in Treg cells.
The O323 antibody may be used for analysis of CD27 expression on peripheral T cells, and is frequently used in combination with antibodies for IgD and IgM to distinguish naïve vs. memory B cell populations. For identification of Breg cells, this antibody has been used in combination with antibodies for CD25, CD1d, IL-10 and TGF-beta (Kessel et al. 2012. Autoimm. Rev. 11(9): 670-677). The antibody is also reported for cross-reactivity with Baboon, Cynomolgus and Rhesus CD27.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: violetFluor™ 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Baboon, Cynomolgus, Rhesus
Specifications
- Catalog No:
- 75-0279-T025
- 75-0279-T100
- Antigen symbol:
- CD27
- CD27
- Antigen name:
- CD27 molecule
- CD27 molecule
- Antigen synonyms:
- Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
- Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- VioletFluor® 450
- VioletFluor® 450
- Clone:
- O323
- O323
- Reactivity:
- Baboon, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 939
- 939
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C...+8 °C, protected from light
- +2 °C...+8 °C, protected from light